Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp2r5d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009358.3 | NP_033384.2 | 15 | 2488 | ||
GenBank mRNA | AB053464.1 | - | 15 | 2434 | ||
AK154050.1 | - | 15 | 2464 | |||
AK204770.1 | - | 1 | 62 | |||
AY338234.1 | - | 15 | 2418 | |||
BC009163.1 | - | 4 | 1149 | |||
BC010716.1 | - | 15 | 2401 | 2655861 | ||
X81059.1 | - | 5 | 528 |