Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tff2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009363.3 | NP_033389.2 | 2 | 188 | ||
GenBank mRNA | AK003093.1 | - | 2 | 189 | ||
BC050086.1 | - | 2 | 149 | |||
BC087889.1 | - | 2 | 166 | 30316277 | ||
X51697.1 | - | 2 | 153 |