Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tgfbi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009369.4 | NP_033395.1 | 17 | 2621 | ||
GenBank mRNA | AK084431.1 | - | 17 | 2639 | ||
AK135536.1 | - | 17 | 2688 | |||
AK142323.1 | - | 17 | 2641 | |||
AK150449.1 | - | 17 | 2639 | |||
AK150833.1 | - | 17 | 2639 | |||
AK150969.1 | - | 17 | 2640 | |||
AK151680.1 | - | 17 | 2636 | |||
AK152365.1 | - | 17 | 2640 | |||
AK152662.1 | - | 17 | 2639 | |||
AK155572.1 | - | 17 | 2636 | |||
AK155828.1 | - | 17 | 2636 | |||
AK159711.1 | - | 17 | 2635 | |||
AK164373.1 | - | 17 | 2640 | |||
AK170495.1 | - | 17 | 2636 | |||
AK171557.1 | - | 16 | 2460 | |||
L19932.1 | - | 17 | 2617 |