Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tgif1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164074.1 | NP_001157546.1 | 3 | 377 | ||
NM_001164075.1 | NP_001157547.1 | 3 | 717 | |||
NM_001164076.1 | NP_001157548.1 | 3 | 443 | |||
NM_001164077.1 | NP_001157549.1 | 2 | 671 | |||
NM_009372.3 | NP_033398.2 | 3 | 631 | |||
GenBank mRNA | AK008909.1 | - | 3 | 632 | ||
AK160193.1 | - | 3 | 336 | |||
BC005724.1 | - | 3 | 630 | 3595273 | ||
BC012700.1 | - | 3 | 728 | 4011149 | ||
X89749.1 | - | 3 | 587 |