Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnfaip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166402.1 | NP_001159874.1 | 5 | 1093 | ||
NM_009397.3 | NP_033423.3 | 6 | 1178 | |||
GenBank mRNA | AK088297.1 | - | 6 | 1135 | ||
AK151921.1 | - | 6 | 1139 | |||
AK153238.1 | - | 6 | 1133 | |||
AK171326.1 | - | 5 | 1092 | |||
AK172529.1 | - | 5 | 1095 | |||
BC050259.1 | - | 5 | 2275 | |||
BC054806.1 | - | 6 | 1178 | 6838303 | ||
BC060221.1 | - | 6 | 1178 | 6843338 | ||
CT010397.1 | - | 5 | 905 | |||
U19463.1 | - | 5 | 905 |