Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009417.2 | NP_033443.1 | 17 | 3056 | ||
GenBank mRNA | AK047843.1 | - | 17 | 3035 | ||
X60703.1 | - | 17 | 3056 |