Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009429.3 | NP_033455.1 | 2, 3 | 277 | ||
GenBank mRNA | AK149695.1 | - | 2, 3 | 203 | ||
AK150300.1 | - | 2, 3 | 204 | |||
AK150500.1 | - | 2, 3 | 203 | |||
AK150858.1 | - | 2, 3 | 204 | |||
AK151059.1 | - | 2, 3 | 205 | |||
AK151266.1 | - | 2, 3 | 203 | |||
AK151375.1 | - | 2, 3 | 203 | |||
AK151894.1 | - | 2, 3 | 203 | |||
AK152690.1 | - | 2, 3 | 203 | |||
AK152808.1 | - | 2, 3 | 204 | |||
AK159516.1 | - | 2, 3 | 203 | |||
AK159565.1 | - | 2, 3 | 203 | |||
AK165736.1 | - | 2, 3 | 203 | |||
AK166584.1 | - | 2, 3 | 206 | |||
AK167413.1 | - | 2, 3 | 203 | |||
AK167936.1 | - | 2, 3 | 203 | |||
AK168151.1 | - | 2, 3 | 203 | |||
AK169730.1 | - | 2, 3 | 205 | |||
AK191401.1 | - | 1, 2 | 34 | |||
BC092381.1 | - | 2, 3 | 163 | 6824238 | ||
X06407.1 | - | 2, 3 | 204 |
Gene Symbol: Arid5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK198991.1 | - | 318 |