Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009442.2 | NP_033468.2 | 2 | 233 | ||
GenBank mRNA | AF237703.1 | - | 1 | 185 | ||
AK140081.1 | - | 2 | 231 | |||
AK163606.1 | - | 2 | 234 | |||
BC059011.1 | - | 1 | 200 | 6847867 | ||
X83974.1 | - | 2, 3 | 219 |