Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRP19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204193.1 | NP_001191122.1 | 5 | 582 | ||
NM_001204194.1 | NP_001191123.1 | 4 | 478 | |||
NM_001204196.1 | NP_001191125.1 | 4 | 474 | |||
NM_003135.2 | NP_003126.1 | 5 | 550 | |||
GenBank mRNA | AK311803.1 | - | 5 | 458 | ||
BC000186.1 | - | 5 | 442 | |||
BC010947.1 | - | 5 | 434 | 4285753 | ||
BC017830.1 | - | 5 | 465 | 4252554 | ||
BU661702.1 | - | 4 | 352 | |||
DN915735.1 | - | 4 | 343 | |||
X12791.1 | - | 5 | 442 |