Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110265.1 | NP_001103735.1 | 5 | 676 | ||
NM_001284272.1 | NP_001271201.1 | 5 | 676 | |||
NM_009445.2 | NP_033471.2 | 5 | 676 | |||
GenBank mRNA | AK041487.1 | - | 5 | 647 | ||
AK049200.1 | - | 5 | 671 | |||
AK164093.1 | - | 5 | 675 | |||
AX775731.1 | - | 5 | 642 | |||
BC058851.1 | - | 5 | 662 | 30023533 | ||
DV648812.1 | - | 2 | 162 | |||
M86377.1 | - | 5 | 624 |