Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ucp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009464.3 | NP_033490.1 | 4 | 619 | ||
GenBank mRNA | AB008216.1 | - | 3 | 422 | ||
AB010742.1 | - | 4 | 619 | |||
AB013132.1 | - | 2 | 175 | |||
AF030164.1 | - | 3 | 460 | |||
AF032902.1 | - | 4 | 586 | |||
AF053352.1 | - | 4 | 591 | |||
BC055901.1 | - | 3 | 809 | |||
BC098234.1 | - | 2 | 1315 | |||
BC139430.1 | - | 3 | 487 | 8862452 | ||
BC139431.1 | - | 3 | 487 | 8862453 |