Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WNT10B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003394.3 | NP_003385.2 | 5 | 1368 | ||
XM_011538722.1 | XP_011537024.1 | 4 | 766 | |||
XM_011538724.1 | XP_011537026.1 | 6 | 1141 | |||
XM_017019919.1 | XP_016875408.1 | 3 | 1400 | |||
GenBank mRNA | AB070724.1 | - | 4 | 972 | ||
AK312906.1 | - | 5 | 1372 | |||
BC096353.3 | - | 5 | 1008 | 40010101 | ||
BC096354.1 | - | 5 | 1008 | 40010102 | ||
BC096355.3 | - | 6 | 866 | |||
BC096356.1 | - | 5 | 1008 | 40010104 | ||
U81787.1 | - | 5 | 1287 |