Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009475.2 | NP_033501.2 | 2, 3 | 148 | ||
GenBank mRNA | AA060392.1 | - | 2, 3 | 146 | ||
BC051092.1 | - | 2, 3 | 136 | 6529097 | ||
U28486.1 | - | 2, 3 | 147 |