Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uty | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009484.2 | NP_033510.2 | 2, 3 | 404 | ||
GenBank mRNA | AF057367.1 | - | 2, 3 | 384 | ||
AK043154.1 | - | 2, 3 | 404 | |||
AK142512.1 | - | 2, 3 | 320 | |||
AK155922.1 | - | 3, 4 | 284 | |||
AK220463.1 | - | 2, 3 | 360 | |||
BC053061.1 | - | 2, 3 | 387 |