Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vmn2r10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009491.3 | NP_033517.3 | 3 | 853 | ||
GenBank mRNA | AF011426.1 | - | 605 |
Set Membership: |
G-protein coupled receptor |