Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vhl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009507.3 | NP_033533.1 | 1, 2 | 366 | ||
GenBank mRNA | AK159238.1 | - | 1, 2 | 387 | ||
AK161408.1 | - | 1, 2 | 363 | |||
AK171264.1 | - | 1, 2 | 368 | |||
AK171604.1 | - | 1, 2 | 376 | |||
BC052417.1 | - | 1, 2 | 365 | 6402536 | ||
S76748.1 | - | 1, 2 | 366 | |||
U12570.1 | - | 1, 2 | 382 |