Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCL2A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114735.1 | NP_001108207.1 | 1 | 479 | ||
NM_004049.3 | NP_004040.1 | 1 | 479 | |||
GenBank mRNA | AL110097.1 | - | 1 | 347 | ||
AY234180.1 | - | 1 | 305 | |||
BC016281.1 | - | 1 | 357 | 3920808 | ||
BG198875.1 | - | 1 | 625 | |||
BG216703.1 | - | 1 | 480 | |||
BT007103.1 | - | 1 | 297 | |||
CD640106.1 | - | 1 | 245 | |||
CR541937.1 | - | 1 | 297 | |||
CR541962.1 | - | 1 | 297 | |||
U27467.1 | - | 1 | 331 | |||
U29680.1 | - | 1 | 375 | |||
Y09397.1 | - | 1 | 399 |