Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wnt6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009526.3 | NP_033552.2 | 4 | 1407 | ||
GenBank mRNA | AK133954.1 | - | 4 | 1408 | ||
BC048700.1 | - | 4 | 1341 | 6511061 | ||
M89800.1 | - | 4 | 1153 |