Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zrsr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009453.3 | NP_033479.2 | 2, 3 | 477 | ||
NM_178794.4 | NP_848909.2 | 2, 3 | 477 | |||
GenBank mRNA | AK030296.1 | - | 2, 3 | 409 | ||
AK045504.1 | - | 2, 3 | 409 | |||
AK078070.1 | - | 2, 3 | 122 | |||
BC065693.1 | - | 2, 3 | 393 | |||
BC083160.1 | - | 2, 3 | 370 | |||
BC099617.1 | - | 2, 3 | 375 | |||
D45205.1 | - | 2, 3 | 400 |