Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2e1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009455.3 | NP_033481.1 | 3 | 361 | ||
GenBank mRNA | AK090177.1 | - | 3 | 362 | ||
AK148266.1 | - | 3 | 292 | |||
BC003781.1 | - | 3 | 278 | 3484294 | ||
X92665.1 | - | 3 | 323 |