Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ulk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009469.3 | NP_033495.2 | 2 | 551 | ||
GenBank mRNA | AF053756.1 | - | 2 | 243 | ||
AF072370.1 | - | 2 | 234 | |||
AK122357.1 | - | 1 | 53 | |||
AK148317.1 | - | 2 | 526 | |||
AK154939.1 | - | 2 | 568 | |||
AK156534.1 | - | 2 | 528 | |||
BC053062.1 | - | 2 | 211 | |||
BC057121.1 | - | 2 | 551 | 6834534 | ||
BC059835.1 | - | 2 | 502 | 6406755 |