Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Umod | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278605.1 | NP_001265534.1 | 3 | 539 | ||
NM_009470.5 | NP_033496.1 | 3 | 595 | |||
GenBank mRNA | AK085460.1 | - | 3 | 589 | ||
AK144065.1 | - | 3 | 511 | |||
AK164688.1 | - | 3 | 511 | |||
AK165507.1 | - | 3 | 455 | |||
BC012973.1 | - | 2 | 433 | 4210940 | ||
L33406.1 | - | 3 | 525 |