Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ezr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009510.2 | NP_033536.2 | 3 | 305 | ||
GenBank mRNA | AK002766.1 | - | 3 | 279 | ||
AK004588.2 | - | 3 | 280 | |||
AK135194.1 | - | 3 | 281 | |||
AK135353.1 | - | 3 | 281 | |||
AK145707.1 | - | 3 | 306 | |||
AK159717.1 | - | 3 | 283 | |||
AK159824.1 | - | 3 | 281 | |||
AK159937.1 | - | 3 | 282 | |||
AK168410.1 | - | 3 | 281 | |||
AK168840.1 | - | 3 | 281 | |||
AK170610.1 | - | 3 | 281 | |||
BC048181.1 | - | 3 | 268 | 6826190 | ||
BC098502.1 | - | 3 | 257 | 30047137 | ||
X60671.1 | - | 3 | 263 |