Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wnt3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009522.2 | NP_033548.1 | 3 | 502 | ||
GenBank mRNA | X56842.1 | - | 3 | 526 |