Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Yap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171147.1 | NP_001164618.1 | 4 | 895 | ||
NM_009534.3 | NP_033560.1 | 4 | 895 | |||
GenBank mRNA | AK182200.1 | - | 2 | 79 | ||
BC014733.1 | - | 4 | 892 | 4239820 | ||
BC039125.1 | - | 4 | 750 | 2649070 | ||
BC094313.1 | - | 4 | 883 | 5697882 | ||
X80508.1 | - | 4 | 887 |