Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009439.1 | NP_033465.1 | 3, 4 | 721 | ||
GenBank mRNA | AK031366.1 | - | 3, 4 | 711 | ||
AK077252.1 | - | 3, 4 | 670 | |||
AK089933.1 | - | 3, 4 | 679 | |||
AK145467.1 | - | 3, 4 | 711 | |||
AK145488.1 | - | 3, 4 | 710 | |||
AK153505.1 | - | 3, 4 | 638 | |||
AK164333.1 | - | 3, 4 | 685 | |||
AK164593.1 | - | 3, 4 | 710 | |||
AK165520.1 | - | 3, 4 | 656 | |||
AK166683.1 | - | 3, 4 | 681 | |||
AK166714.1 | - | 3, 4 | 709 | |||
AK167005.1 | - | 3, 4 | 696 | |||
AK167285.1 | - | 3, 4 | 676 | |||
AK169690.1 | - | 3, 4 | 710 | |||
AK180273.1 | - | 1, 2 | 97 | |||
BC003197.1 | - | 3, 4 | 721 | 3586427 |