Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009442.2 | NP_033468.2 | 10 | 2378 | ||
GenBank mRNA | AF237703.1 | - | 9 | 2330 | ||
AK140081.1 | - | 9 | 3596 | |||
BC059011.1 | - | 9 | 2345 | 6847867 | ||
X83974.1 | - | 11 | 2286 |