Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uba1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136085.2 | NP_001129557.1 | 5 | 598 | ||
NM_001276316.1 | NP_001263245.1 | 6 | 690 | |||
NM_001276317.1 | NP_001263246.1 | 5 | 593 | |||
NM_009457.4 | NP_033483.2 | 5 | 593 | |||
GenBank mRNA | AK081808.1 | - | 5 | 564 | ||
AK088057.1 | - | 5 | 475 | |||
AK088528.1 | - | 5 | 475 | |||
AK143416.1 | - | 5 | 499 | |||
AK171667.1 | - | 5 | 564 | |||
AK194691.1 | - | 2 | 128 | |||
AK197186.1 | - | 2 | 128 | |||
AK197943.1 | - | 2 | 128 | |||
AK199667.1 | - | 2 | 128 | |||
AK205902.1 | - | 2 | 128 | |||
AK209874.1 | - | 2 | 128 | |||
AK210948.1 | - | 2 | 128 | |||
AK215149.1 | - | 2 | 128 | |||
AK215450.1 | - | 2 | 128 | |||
AK219140.1 | - | 2 | 128 | |||
AK219193.1 | - | 2 | 128 | |||
BC058630.1 | - | 5 | 539 | 6402368 | ||
BC138200.1 | - | 5 | 456 | 8861219 | ||
BC145984.1 | - | 5 | 457 | 40131287 | ||
D10576.1 | - | 5 | 459 |