Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sumo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009460.2 | NP_033486.1 | 5 | 1054 | ||
GenBank mRNA | AF033353.1 | - | 5 | 1029 | ||
AK002536.1 | - | 5 | 1024 | |||
AK011074.1 | - | 5 | 1023 | |||
AK012235.1 | - | 1038 | ||||
AK089081.1 | - | 5 | 1003 | |||
AK159366.1 | - | 5 | 1025 | |||
BC082566.1 | - | 5 | 1029 | 30602751 | ||
BC083158.1 | - | 5 | 1027 | 6478032 |