Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCEB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204857.1 | NP_001191786.1 | 3, 4 | 249 | ||
NM_001204858.1 | NP_001191787.1 | 4, 5 | 560 | |||
NM_001204859.1 | NP_001191788.1 | 4, 5 | 556 | |||
NM_001204860.1 | NP_001191789.1 | 4, 5 | 388 | |||
NM_001204861.1 | NP_001191790.1 | 5, 6 | 440 | |||
NM_001204862.1 | NP_001191791.1 | 4, 5 | 301 | |||
NM_001204863.1 | NP_001191792.1 | 2, 3 | 363 | |||
NM_001204864.1 | NP_001191793.1 | 2, 3 | 195 | |||
NM_005648.3 | NP_005639.1 | 3, 4 | 417 | |||
XM_011517580.2 | XP_011515882.1 | 3, 4 | 315 | |||
XM_011517581.2 | XP_011515883.1 | 3, 4 | 325 | |||
GenBank mRNA | AK057889.1 | - | 3, 4 | 297 | ||
AL832244.1 | - | 3, 4 | 250 | |||
BC013809.1 | - | 3, 4 | 241 | 3923736 | ||
BC093065.1 | - | 3, 4 | 248 | 30411791 | ||
BC100028.1 | - | 3, 4 | 266 | 6062030 | ||
BC100283.1 | - | 3, 4 | 377 | 6061521 | ||
BG428202.1 | - | 3, 4 | 249 | |||
BG703267.1 | - | 2, 3 | 338 | |||
BM720777.1 | - | 5, 6 | 441 | |||
BM821650.1 | - | 3, 4 | 400 | |||
BX649138.1 | - | 2, 3 | 247 | |||
CF129635.1 | - | 2, 3 | 182 | |||
CX166168.1 | - | 4, 5 | 285 | |||
CX166843.1 | - | 3, 4 | 356 | |||
DA537957.1 | - | 4, 5 | 390 | |||
L34587.1 | - | 2, 3 | 243 |