Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nr1h2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285517.1 | NP_001272446.1 | 8 | 1261 | ||
NM_001285518.1 | NP_001272447.1 | 8 | 1255 | |||
NM_001285519.1 | NP_001272448.1 | 8 | 1252 | |||
NM_009473.3 | NP_033499.1 | 8 | 1268 | |||
GenBank mRNA | AK077620.1 | - | 8 | 1234 | ||
AK134489.1 | - | 8 | 1218 | |||
AK156777.1 | - | 8 | 1236 | |||
AK177488.1 | - | 1 | 19 | |||
AK194055.1 | - | 1 | 19 | |||
BC066025.1 | - | 8 | 1219 | 6829527 | ||
BC106148.1 | - | 7 | 988 | |||
EU869275.2 | - | 5 | 1247 | |||
U09419.1 | - | 7 | 1086 |