Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp9x | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009481.2 | NP_033507.2 | 35 | 6294 | ||
GenBank mRNA | AK034941.1 | - | 1 | 355 | ||
BY716634.1 | - | 1 | 397 | |||
DQ086491.1 | - | 35 | 6255 | |||
U67874.1 | - | 35 | 6255 |