Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTDSP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005730.3 | NP_005721.3 | 2 | 726 | ||
XM_005268556.1 | XP_005268613.1 | 2 | 486 | |||
GenBank mRNA | AF000152.1 | - | 2 | 502 | ||
AF022231.1 | - | 2 | 443 | |||
AK226076.1 | - | 2 | 482 | |||
AK291289.1 | - | 2 | 473 | |||
AK310873.1 | - | 2 | 228 | |||
AK312413.1 | - | 2 | 485 | |||
AY279531.1 | - | 2 | 197 | |||
BC018922.2 | - | 2 | 458 | |||
BC065920.1 | - | 2 | 441 | 5756196 | ||
CN313995.1 | - | 2 | 651 |