Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vcp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009503.4 | NP_033529.3 | 4, 5 | 782 | ||
GenBank mRNA | AK028264.1 | - | 4, 5 | 711 | ||
AK030751.1 | - | 4, 5 | 613 | |||
AK083821.1 | - | 2, 3 | 288 | |||
AK146109.1 | - | 4, 5 | 679 | |||
AK149931.1 | - | 4, 5 | 675 | |||
AK151109.1 | - | 4, 5 | 676 | |||
AK151418.1 | - | 4, 5 | 676 | |||
AK153012.1 | - | 4, 5 | 712 | |||
AK153249.1 | - | 4, 5 | 753 | |||
AK159177.1 | - | 4, 5 | 711 | |||
AK159475.1 | - | 4, 5 | 710 | |||
AK159509.1 | - | 4, 5 | 754 | |||
AK160889.1 | - | 4, 5 | 599 | |||
AK167794.1 | - | 4, 5 | 730 | |||
AK169140.1 | - | 4, 5 | 684 | |||
AK183217.1 | - | 1, 2 | 117 | |||
AK185530.1 | - | 1, 2 | 117 | |||
AK195731.1 | - | 1, 2 | 109 | |||
AK198927.1 | - | 1, 2 | 109 | |||
AK213510.1 | - | 1, 2 | 109 | |||
AK213824.1 | - | 1, 2 | 109 | |||
AK216568.1 | - | 1, 2 | 109 | |||
BC043053.1 | - | 4, 5 | 686 | 6418662 | ||
BC049114.1 | - | 4, 5 | 730 | 6810751 | ||
Z14044.1 | - | 4, 5 | 614 |