Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MANF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006010.5 | NP_006001.4 | 2 | 319 | ||
GenBank mRNA | AK292243.1 | - | 2 | 232 | ||
BC007282.1 | - | 2 | 215 | |||
BC113588.1 | - | 2 | 242 | 8322640 | ||
BC113590.1 | - | 2 | 242 | 8322642 | ||
BE514267.1 | - | 2 | 326 | |||
BT007110.1 | - | 2 | 213 | |||
CN410911.1 | - | 2 | 308 | |||
M83751.1 | - | 2 | 501 |