Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp185 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109043.1 | NP_001102513.1 | 10, 11 | 868 | ||
NM_009549.3 | NP_033575.3 | 10, 11 | 871 | |||
GenBank mRNA | AK048188.1 | - | 10, 11 | 849 | ||
BC039772.1 | - | 7, 8 | 456 | 4191053 | ||
EF494744.1 | - | 10, 11 | 851 | |||
U46687.1 | - | 9, 10 | 697 |