Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp94 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199321.1 | NP_001186250.1 | 6 | 515 | ||
NM_009568.3 | NP_033594.1 | 5 | 476 | |||
GenBank mRNA | AK138205.1 | - | 6 | 456 | ||
AK164334.1 | - | 6 | 468 | |||
BC005437.1 | - | 5 | 476 | 3591835 | ||
BC095945.1 | - | 4 | 296 | 4936327 | ||
CJ174209.1 | - | 5 | 378 | |||
U46187.1 | - | 5 | 419 |