Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zhx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042438.2 | NP_001035903.1 | 4 | 1169 | ||
NM_009572.4 | NP_033598.2 | 3 | 1043 | |||
GenBank mRNA | AK051410.1 | - | 3 | 1032 | ||
AK053191.1 | - | 3 | 753 | |||
AK220508.1 | - | 3 | 1004 | |||
BC004723.1 | - | 4 | 875 | |||
BC054543.1 | - | 2 | 627 | 6406286 | ||
Z54200.1 | - | 3 | 754 |