Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zik1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009577.3 | NP_033603.2 | 3, 4 | 413 | ||
GenBank mRNA | AK146692.1 | - | 3, 4 | 407 | ||
AK159077.1 | - | 3, 4 | 399 | |||
BC050942.1 | - | 3, 4 | 371 | 6821378 | ||
BC067196.1 | - | 3, 4 | 371 | 6821378 | ||
U69133.1 | - | 3, 4 | 411 |