Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc30a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009579.3 | NP_033605.1 | 1, 2 | 837 | ||
GenBank mRNA | AK145100.1 | - | 1, 2 | 862 | ||
BC052166.1 | - | 1, 2 | 828 | 6333155 |