Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map3k12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163643.1 | NP_001157115.1 | 4 | 504 | ||
NM_009582.4 | NP_033608.3 | 2 | 328 | |||
GenBank mRNA | AK029882.1 | - | 2 | 137 | ||
AK034374.1 | - | 4 | 320 | |||
AK166435.1 | - | 4 | 424 | |||
BC047158.1 | - | 1 | 122 | 6494216 | ||
BC057572.1 | - | 4 | 328 | 6401321 | ||
U14636.1 | - | 2 | 160 |