Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ache | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290010.1 | NP_001276939.1 | 2 | 412 | ||
NM_009599.4 | NP_033729.1 | 2 | 409 | |||
GenBank mRNA | AK036259.1 | - | 2 | 383 | ||
AK036443.1 | - | 1 | 2023 | |||
AK042335.1 | - | 2 | 387 | |||
AK043748.1 | - | 2 | 383 | |||
AK046080.1 | - | 2 | 380 | |||
AK140404.1 | - | 2 | 383 | |||
BC046327.1 | - | 1 | 377 | 6468980 | ||
EU623420.1 | - | 1 | 273 | |||
X56518.1 | - | 1 | 273 |