Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chrne | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009603.1 | NP_033733.1 | 2 | 189 | ||
GenBank mRNA | AF032863.1 | - | 1 | 42 | ||
X55718.1 | - | 2 | 189 |