Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ak4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177602.1 | NP_001171073.1 | 2 | 439 | ||
NM_001177604.1 | NP_001171075.1 | 2 | 252 | |||
NM_001177605.1 | NP_001171076.1 | 2 | 230 | |||
NM_009647.5 | NP_033777.1 | 1 | 197 | |||
GenBank mRNA | AB020239.1 | - | 1 | 161 | ||
AK134600.1 | - | 2 | 442 | |||
AK144439.1 | - | 1 | 167 | |||
AK154374.1 | - | 2 | 256 | |||
AK166975.1 | - | 1 | 119 | |||
BC086663.1 | - | 1 | 145 | 30649610 | ||
BC086791.1 | - | 1 | 161 | |||
BY718404.1 | - | 2 | 440 | |||
CJ087385.1 | - | 1 | 197 | |||
D85036.1 | - | 1 | 161 |