Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ikzf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK036833.1 | - | 7 | 989 | ||
AK040268.1 | - | 6 | 722 | |||
AK077815.1 | - | 7 | 1286 | |||
AK142856.1 | - | 5 | 486 | |||
AK154175.1 | - | 6 | 3913 | |||
AK220235.1 | - | 7 | 955 | |||
BC138789.1 | - | 6 | 520 | 8861811 | ||
L03547.1 | - | 6 | 505 | |||
S74708.1 | - | 4 | 566 |