Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chrnb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009601.4 | NP_033731.3 | 3 | 273 | ||
GenBank mRNA | AK087554.1 | - | 3 | 252 | ||
BC023318.1 | - | 3 | 279 | 4009493 | ||
CJ176855.1 | - | 3 | 273 | |||
M14537.1 | - | 3 | 216 |