Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chrnb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009602.4 | NP_033732.2 | 4 | 525 | ||
GenBank mRNA | AF145286.1 | - | 4 | 464 | ||
AF299083.1 | - | 4 | 319 | |||
AF325349.1 | - | 4 | 285 | |||
AK049722.1 | - | 4 | 498 | |||
AK051742.1 | - | 4 | 507 | |||
AK081254.1 | - | 502 | ||||
AY574267.1 | - | 4 | 299 | |||
BC065103.1 | - | 4 | 478 | 6838257 |