Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chrng | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009604.3 | NP_033734.3 | 4 | 324 | ||
GenBank mRNA | AK047974.1 | - | 4 | 325 | ||
JF520813.1 | - | 4 | 325 | |||
JN164665.1 | - | 492 | ||||
JN164666.1 | - | 3 | 383 | |||
M30514.1 | - | 4 | 365 | |||
X03818.1 | - | 4 | 321 | |||
X03819.1 | - | 3 | 256 |