Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acvrl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277255.1 | NP_001264184.1 | 6 | 795 | ||
NM_001277257.1 | NP_001264186.1 | 5 | 734 | |||
NM_001277258.1 | NP_001264187.1 | 5 | 703 | |||
NM_001277259.1 | NP_001264188.1 | 6 | 790 | |||
NM_009612.3 | NP_033742.2 | 5 | 772 | |||
GenBank mRNA | AK018754.1 | - | 2 | 274 | ||
AK134712.1 | - | 3 | 529 | |||
AK160915.1 | - | 6 | 763 | |||
AK170870.1 | - | 3 | 528 | |||
BC014291.1 | - | 4 | 691 | 3982004 | ||
BC015083.1 | - | 4 | 653 | 3980151 | ||
CT010330.1 | - | 3 | 500 | |||
L48015.1 | - | 5 | 774 | |||
Z31664.1 | - | 5 | 740 |