Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adamts1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009621.4 | NP_033751.3 | 1 | 896 | ||
GenBank mRNA | AK156830.1 | - | 1 | 895 | ||
AK163622.1 | - | 1 | 895 | |||
BC003269.1 | - | 1 | 881 | |||
BC040382.1 | - | 1 | 880 | 3156736 | ||
BC050834.1 | - | 1 | 880 | 6505494 | ||
D67076.1 | - | 1 | 880 |